Assay Details
| Gene Symbol: Ccdc127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198766.1 | NP_942061.1 | 1 - 2 | 85 | 105 | |
| XM_006227783.2 | XP_006227845.1 | 1 - 2 | 105 | |||
| GenBank mRNA | BC058157.1 | - | 1 - 2 | 85 | 6890640 | 105 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058157.1, NM_198766.1, XM_006227783.2 transcripts.
Set Membership: |
|

Ccdc127
Genomic Map