Assay Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100735.1 | NP_001094205.1 | 9 - 10 | 1035 | 75 | |
| GenBank mRNA | AB244983.2 | - | 8 - 9 | 763 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid biosynthetic process
phosphatidylcholine acyl-chain remodeling
surfactant homeostasis
positive regulation of protein catabolic process
retina development in camera-type eye
negative regulation of phosphatidylcholine biosynthetic process
calcium ion binding
1-alkenylglycerophosphocholine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
phospholipid biosynthetic process
phosphatidylcholine acyl-chain remodeling
surfactant homeostasis
positive regulation of protein catabolic process
retina development in camera-type eye
negative regulation of phosphatidylcholine biosynthetic process
calcium ion binding
1-alkenylglycerophosphocholine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P942245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25362 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P942245 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P942245 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P942245 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P942245 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P178631 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102439 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10717575 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717585 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717572 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717577 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717582 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717578 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717574 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717584 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717583 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717581 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717576 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717579 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717573 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717580 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10717571 | - | - |
| Rat Genome 230 2.0 Array | 1371923_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371923_at | - | - |
| Affymetrix HT Rat Focus Array | 1371923_at | - | - |
| Affymetrix HT Rat Focus Array | 1371923_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100034292.rn.1 | TC0100004280.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371923_at | - | - |
| Rat Expression Set 230 | 1371923_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10717571 | - | - |
| Rat Genome U34 Set | rc_AA818832_at | - | - |
| Rat Genome U34 Set | rc_AI228324_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22400280 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357096 | - | - |

Lpcat1
Genomic Map