Assay Details
| Gene Symbol: Anxa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031654.1 | NP_001026824.1 | 7 - 8 | 565 | 84 | |
| GenBank mRNA | BC099106.1 | - | 7 - 8 | 565 | 7460531 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Anxa8
Genomic Map