Assay Details
| Gene Symbol: Ptpn20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080149.1 | NP_001073618.1 | 1 - 2 | 82 | 97 | |
| XM_006252765.2 | XP_006252827.1 | 2 - 3 | 155 | 97 | ||
| XM_008771099.1 | XP_008769321.1 | 2 - 3 | 108 | 97 | ||
| XM_008771100.1 | XP_008769322.1 | 2 - 3 | 155 | 97 | ||
| GenBank mRNA | BC129117.1 | - | 1 - 2 | 82 | 7114504 | 97 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC129117.1,NM_001080149.1,XM_006252765.2,XM_008771099.1,XM_008771100.1 transcripts.
Set Membership: |
|

Ptpn20b
Genomic Map