Assay Details
| Gene Symbol: LOC685179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001055673.5 | XP_001055673.1 | 23 - 24 | 2472 | 168 | |
| XM_001055738.5 | XP_001055738.1 | 22 - 23 | 2379 | 168 | ||
| XM_001055795.5 | XP_001055795.1 | 22 - 23 | 2380 | 168 | ||
| XM_002729767.4 | XP_002729813.2 | 23 - 24 | 2473 | 168 | ||
| XM_006240805.2 | XP_006240867.1 | 23 - 24 | 2470 | 168 | ||
| XM_006240806.2 | XP_006240868.1 | 23 - 24 | 2452 | 168 | ||
| XM_006240807.2 | XP_006240869.1 | 22 - 23 | 2377 | 168 | ||
| XM_006240808.2 | XP_006240870.1 | 22 - 23 | 2359 | 168 | ||
| XM_008765050.1 | XP_008763272.1 | 23 - 24 | 2473 | 168 | ||
| XM_008765051.1 | XP_008763273.1 | 23 - 24 | 3317 | 168 | ||
More Information
Set Membership: |
|

LOC685179
Genomic Map