Assay Details
| Gene Symbol: LOC685179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001055673.5 | XP_001055673.1 | 25 - 26 | 2775 | 118 | |
| XM_001055738.5 | XP_001055738.1 | 24 - 25 | 2682 | 118 | ||
| XM_001055795.5 | XP_001055795.1 | 24 - 25 | 2683 | 118 | ||
| XM_002729767.4 | XP_002729813.2 | 25 - 26 | 2776 | 118 | ||
| XM_006240805.2 | XP_006240867.1 | 25 - 26 | 2773 | 118 | ||
| XM_006240806.2 | XP_006240868.1 | 25 - 26 | 2755 | 118 | ||
| XM_006240807.2 | XP_006240869.1 | 24 - 25 | 2680 | 118 | ||
| XM_006240808.2 | XP_006240870.1 | 24 - 25 | 2662 | 118 | ||
| XM_008765050.1 | XP_008763272.1 | 25 - 26 | 2776 | 118 | ||
| XM_008765051.1 | XP_008763273.1 | 25 - 26 | 3620 | 118 | ||
More Information
Set Membership: |
|

LOC685179
Genomic Map