Assay Details
| Gene Symbol: Elk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108059.3 | NP_001101529.1 | 4 - 5 | 853 | 107 | |
| XM_006256614.2 | XP_006256676.1 | 3 - 4 | 1835 | 107 | ||
| GenBank mRNA | EF429098.1 | - | 2 - 3 | 651 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elk1
Genomic Map