Assay Details
| Gene Symbol: Alg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108401.1 | NP_001101871.1 | 2 - 3 | 287 | 110 | |
| XM_006253354.2 | XP_006253416.1 | 2 - 3 | 559 | 110 | ||
| XM_006253355.2 | XP_006253417.1 | 2 - 3 | 261 | 110 | ||
| XM_006253356.2 | XP_006253418.1 | 2 - 3 | 884 | 110 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Alg11
Genomic Map