Assay Details
| Gene Symbol: Proz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253473.1 | XP_006253535.1 | 2 - 3 | 349 | 72 | |
| GenBank mRNA | BC160868.1 | - | 2 - 3 | 261 | 72 | |
| FQ209855.1 | - | 2 - 3 | 263 | 72 | ||
More Information
Set Membership: |
|

Proz
Genomic Map