Assay Details
| Gene Symbol: Slc25a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047880.1 | NP_001041345.1 | 3 - 4 | 353 | 62 | |
| XM_006253350.2 | XP_006253412.1 | 3 - 4 | 789 | 62 | ||
| XM_006253351.2 | XP_006253413.1 | 3 - 4 | 1180 | 62 | ||
| XM_006253352.2 | XP_006253414.1 | 3 - 4 | 1006 | 62 | ||
| XM_006253353.2 | XP_006253415.1 | 2 - 3 | 1834 | 62 | ||
| GenBank mRNA | AY325139.1 | - | 3 - 4 | 353 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc25a15
Genomic Map