Assay Details
| Gene Symbol: LOC100363275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751617.3 | XP_003751665.2 | 9 - 10 | 1723 | 69 | |
| XM_006253295.2 | XP_006253357.1 | 9 - 10 | 1723 | 69 | ||
More Information
Set Membership: |
|

LOC100363275
Genomic Map