Assay Details
| Gene Symbol: Cnpy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134710.1 | NP_001128182.1 | 1 - 2 | 167 | 60 | |
| GenBank mRNA | BC166762.1 | - | 1 - 2 | 167 | 7931606 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Cnpy3
Genomic Map