Assay Details
| Gene Symbol: Tnfrsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108207.1 | NP_001101677.1 | 1 - 2 | 544 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf21| Gene Symbol: Tnfrsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108207.1 | NP_001101677.1 | 1 - 2 | 544 | 72 | |
Set Membership: |
|