Assay Details
| Gene Symbol: Supt3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244587.2 | XP_006244649.2 | 5 - 6 | 1136 | 84 | |
| XM_008766906.1 | XP_008765128.1 | 5 - 6 | 1136 | 84 | ||
| XM_008766907.1 | XP_008765129.1 | 5 - 6 | 609 | 84 | ||
| XM_008766908.1 | XP_008765130.1 | 5 - 6 | 505 | 84 | ||
| XM_008766909.1 | XP_008765131.1 | 5 - 6 | 466 | 84 | ||
| XM_008766910.1 | XP_008765132.1 | 4 - 5 | 379 | 84 | ||
| XM_008766911.1 | XP_008765133.1 | 4 - 5 | 364 | 84 | ||
More Information
Set Membership: |
|

Supt3h
Genomic Map