Assay Details
| Gene Symbol: LOC100911260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229638.1 | XP_006229700.1 | 5 - 6 | 706 | 74 | |
| Gene Symbol: Hcfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139507.1 | NP_001132979.1 | 16 - 17 | 3975 | 74 | |
| XM_006229593.2 | XP_006229655.1 | 16 - 17 | 3954 | 74 | ||
| XM_006229594.2 | XP_006229656.1 | 16 - 17 | 3953 | 74 | ||
| XM_006229596.2 | XP_006229658.1 | 16 - 17 | 3951 | 74 | ||
| XM_006229597.1 | XP_006229659.1 | 16 - 17 | 3970 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911260
Genomic Map