Assay Details
| Gene Symbol: LOC100911260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229638.1 | XP_006229700.1 | 6 - 7 | 2212 | 67 | |
| Gene Symbol: Hcfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139507.1 | NP_001132979.1 | 17 - 18 | 5481 | 67 | |
| XM_006229593.2 | XP_006229655.1 | 17 - 18 | 5460 | 67 | ||
| XM_006229594.2 | XP_006229656.1 | 17 - 18 | 5459 | 67 | ||
| XM_006229596.2 | XP_006229658.1 | 17 - 18 | 5457 | 67 | ||
| XM_006229597.1 | XP_006229659.1 | 17 - 18 | 5476 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911260
Genomic Map