Assay Details
| Gene Symbol: LOC100911260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229638.1 | XP_006229700.1 | 9 - 10 | 3275 | 63 | |
| Gene Symbol: Hcfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139507.1 | NP_001132979.1 | 20 - 21 | 6412 | 63 | |
| XM_006229593.2 | XP_006229655.1 | 20 - 21 | 6526 | 63 | ||
| XM_006229594.2 | XP_006229656.1 | 20 - 21 | 6522 | 63 | ||
| XM_006229596.2 | XP_006229658.1 | 20 - 21 | 6391 | 63 | ||
| XM_006229597.1 | XP_006229659.1 | 20 - 21 | 6407 | 63 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911260
Genomic Map