Assay Details
| Gene Symbol: LOC102556049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253911.1 | XP_006253973.1 | 5 - 6 | 566 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102556049| Gene Symbol: LOC102556049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253911.1 | XP_006253973.1 | 5 - 6 | 566 | 133 | |
Set Membership: |
|