Assay Details
Gene Symbol: Rnf144b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108881.1 | NP_001102351.1 | 3 - 4 | 454 | 70 | |
XM_006253767.2 | XP_006253829.1 | 3 - 4 | 887 | 70 | ||
XM_008771549.1 | XP_008769771.1 | 3 - 4 | 892 | 70 | ||
XM_008771550.1 | XP_008769772.1 | 3 - 4 | 980 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
