Assay Details
| Gene Symbol: Ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107339.1 | NP_001100809.1 | 8 - 9 | 1590 | 70 | |
| XM_008771495.1 | XP_008769717.1 | 7 - 8 | 1302 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
cartilage condensation
somitogenesis
transmembrane receptor protein tyrosine kinase signaling pathway
Wnt signaling pathway, calcium modulating pathway
smoothened signaling pathway
JNK cascade
negative regulation of cell proliferation
peptidyl-tyrosine phosphorylation
cell differentiation
positive regulation of cell migration
BMP signaling pathway
embryonic genitalia morphogenesis
inner ear morphogenesis
embryonic digit morphogenesis
cell fate commitment
positive regulation of transcription, DNA-templated
Wnt signaling pathway, planar cell polarity pathway
SMAD protein signal transduction
regulation of canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
transmembrane receptor protein tyrosine kinase activity
frizzled binding
ATP binding
Wnt-protein binding
skeletal system development
cartilage condensation
somitogenesis
transmembrane receptor protein tyrosine kinase signaling pathway
Wnt signaling pathway, calcium modulating pathway
smoothened signaling pathway
JNK cascade
negative regulation of cell proliferation
peptidyl-tyrosine phosphorylation
cell differentiation
positive regulation of cell migration
BMP signaling pathway
embryonic genitalia morphogenesis
inner ear morphogenesis
embryonic digit morphogenesis
cell fate commitment
positive regulation of transcription, DNA-templated
Wnt signaling pathway, planar cell polarity pathway
SMAD protein signal transduction
regulation of canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
transmembrane receptor protein tyrosine kinase activity
frizzled binding
ATP binding
Wnt-protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P153956 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P153952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115558 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153951 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153956 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153956 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153951 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P153951 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P153956 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19594 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43135 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30933 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97860 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74204 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10797502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797505 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10797499 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700001733.rn.1 | TC1700000209.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10797499 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21521118 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372250 | - | - |

Ror2
Genomic Map