Assay Details
| Gene Symbol: Prl8a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020079.1 | NP_064464.1 | 392 | 87 | ||
| GenBank mRNA | U93351.1 | - | 392 | 87 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P942715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28372 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P942715 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P942715 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P942715 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P942715 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61792 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62994 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6266 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10795052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795053 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795055 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795051 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795054 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10795050 | - | - |
| Rat Genome 230 2.0 Array | 1368531_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368531_at | - | - |
| Affymetrix HT Rat Focus Array | 1368531_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368531_at | - | - |
| Rat Expression Set 230 | 1368531_at | - | - |
| Rat Expression Set 230 | 1368531_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10795050 | - | - |
| Rat Genome U34 Set | rc_AA894246_f_at | - | - |
| Rat Genome U34 Set | rc_AI180410_f_at | - | - |
| Rat Genome U34 Set | rc_AI180410_r_at | - | - |
| Rat Genome U34 Set | rc_AI180410_i_at | - | - |
| Rat Genome U34 Set | rc_AI180452_f_at | - | - |
| Rat Genome U34 Set | rc_AI180452_i_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21911208 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370421 | - | - |
| RatRef-12 Expression BeadChip | ILMN_67888 | - | - |

Prl8a3
Genomic Map