Assay Details
| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752076.3 | XP_003752124.1 | 3 - 4 | 702 | 75 | |
| XM_008773325.1 | XP_008771547.1 | 2 - 3 | 2331 | 75 | ||
More Information
Set Membership: |
|

Zfp449
Genomic Map