Assay Details
| Gene Symbol: Zfp449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752076.3 | XP_003752124.1 | 4 - 5 | 819 | 149 | |
| XM_008773325.1 | XP_008771547.1 | 3 - 4 | 2448 | 149 | ||
More Information
Set Membership: |
|

Zfp449
Genomic Map