Assay Details
| Gene Symbol: Tox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106171.1 | NP_001099641.1 | 5 - 6 | 903 | 57 | |
| XM_006255187.2 | XP_006255249.1 | 6 - 7 | 1052 | 57 | ||
| XM_006255189.2 | XP_006255251.1 | 6 - 7 | 1049 | 57 | ||
| GenBank mRNA | EU194254.1 | - | 6 - 7 | 991 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
apoptotic process
calcium-mediated signaling
negative regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
chromatin binding
protein binding
estrogen response element binding
protein homodimerization activity
phosphoprotein binding
chromatin remodeling
transcription, DNA-templated
apoptotic process
calcium-mediated signaling
negative regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
chromatin binding
protein binding
estrogen response element binding
protein homodimerization activity
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P632469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43206 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P632469 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P632469 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P632469 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P141767 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14741 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10806306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806309 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806308 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806310 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806307 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10806303 | - | - |
| Rat Genome 230 2.0 Array | 1379751_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379751_at | - | - |
| Rat Genome 230 2.0 Array | 1377582_at | - | - |
| Rat Genome 230 2.0 Array | 1382579_at | - | - |
| Rat Genome 230 2.0 Array | 1382579_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377582_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382579_at | - | - |
| Affymetrix HT Rat Focus Array | 1382579_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379751_at | - | - |
| Affymetrix HT Rat Focus Array | 1379751_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900001544.rn.1 | TC1900000198.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900001543.rn.1 | TC1900000198.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900001545.rn.1 | TC1900000198.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900001546.rn.1 | TC1900000198.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382579_PM_at | - | - |
| Rat Expression Set 230 | 1377582_at | - | - |
| Rat Expression Set 230 | 1377582_PM_at | - | - |
| Rat Expression Set 230 | 1379751_at | - | - |
| Rat Expression Set 230 | 1379751_PM_at | - | - |
| Rat Expression Set 230 | 1382579_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10806303 | - | - |
| Rat Genome U34 Set | rc_AA997563_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21335514 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367193 | - | - |

Tox3
Genomic Map