Assay Details
| Gene Symbol: Naip6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231862.2 | XP_006231924.1 | 15 - 16 | 4385 | 63 | |
| XM_006231863.2 | XP_006231925.1 | 15 - 16 | 4189 | 63 | ||
| XM_008760695.1 | XP_008758917.1 | 19 - 20 | 4736 | 63 | ||
| XM_008760696.1 | XP_008758918.1 | 18 - 19 | 4631 | 63 | ||
| XM_008760697.1 | XP_008758919.1 | 18 - 19 | 4617 | 63 | ||
| XM_008760698.1 | XP_008758920.1 | 14 - 15 | 4051 | 63 | ||
| GenBank mRNA | FQ234105.1 | - | 2 - 3 | 242 | 63 | |
More Information
Set Membership: |
|

Naip6
Genomic Map