Assay Details
| Gene Symbol: RGD1562101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760641.1 | XP_008758863.1 | 20 - 21 | 4600 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562101| Gene Symbol: RGD1562101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760641.1 | XP_008758863.1 | 20 - 21 | 4600 | 74 | |
Set Membership: |
|