Assay Details
| Gene Symbol: Gpr98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D87256.1 | - | 7 - 8 | 1329 | 110 | |
| Gene Symbol: RGD1562101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760641.1 | XP_008758863.1 | 7 - 8 | 1470 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr98
Genomic Map