Assay Details
| Gene Symbol: Mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191605.3 | NP_001178534.1 | 3 - 4 | 374 | 65 | |
| XM_006231846.2 | XP_006231908.1 | 3 - 4 | 498 | 65 | ||
| GenBank mRNA | FM072467.1 | - | 3 - 4 | 374 | 65 | |
| FQ224224.1 | - | 3 - 4 | 308 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps36
Genomic Map