Assay Details
| Gene Symbol: Rhobtb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107645.1 | NP_001101115.1 | 2 - 3 | 650 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhobtb3| Gene Symbol: Rhobtb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107645.1 | NP_001101115.1 | 2 - 3 | 650 | 58 | |
Set Membership: |
|