Assay Details
| Gene Symbol: Aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022851.1 | NP_074042.1 | 8 - 9 | 1321 | 70 | |
| GenBank mRNA | L34821.1 | - | 8 - 9 | 1200 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh5a1
Genomic Map