Assay Details
| Gene Symbol: Gfi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107823.1 | NP_001101293.1 | 4 - 5 | 626 | 77 | |
| XM_006233789.1 | XP_006233851.1 | 4 - 5 | 665 | 77 | ||
| XM_008761646.1 | XP_008759868.1 | 4 - 5 | 713 | 77 | ||
| XM_008761647.1 | XP_008759869.1 | 4 - 5 | 3987 | 77 | ||
| XM_008761648.1 | XP_008759870.1 | 5 - 6 | 877 | 77 | ||
| XM_008761649.1 | XP_008759871.1 | 4 - 5 | 787 | 77 | ||
| XM_008761650.1 | XP_008759872.1 | 4 - 5 | 1265 | 77 | ||
| XM_008761651.1 | XP_008759873.1 | 4 - 5 | 713 | 77 | ||
| GenBank mRNA | BC158685.1 | - | 4 - 5 | 641 | 8374780 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Gfi1b
Genomic Map