Assay Details
| Gene Symbol: Ssna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233613.1 | XP_006233675.1 | 1 - 2 | 147 | 64 | |
| GenBank mRNA | BC166994.1 | - | 1 - 2 | 63 | 7304458 | 64 |
| CB576781.1 | - | 1 - 2 | 100 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssna1
Genomic Map