Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100532.1 | NP_001094002.1 | 2 - 3 | 787 | 60 | |
| GenBank mRNA | BC166796.1 | - | 2 - 3 | 787 | 7938000 | 60 |
| FQ224461.1 | - | 2 - 3 | 797 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map