Assay Details
| Gene Symbol: Tbx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107193.1 | NP_001100663.1 | 2 - 3 | 670 | 94 | |
| XM_008769651.1 | XP_008767873.1 | 2 - 3 | 1346 | 94 | ||
| XM_008769652.1 | XP_008767874.1 | 3 - 4 | 707 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbx19
Genomic Map