Assay Details
| Gene Symbol: Mr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100635.1 | NP_001094105.1 | 1 - 2 | 50 | 77 | |
| XM_006250030.2 | XP_006250092.1 | 1 - 2 | 164 | 77 | ||
| XM_008769606.1 | XP_008767828.1 | 1 - 2 | 164 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mr1
Genomic Map