Assay Details
| Gene Symbol: Lmx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105967.2 | NP_001099437.2 | 3 - 4 | 710 | 86 | |
| XM_008769728.1 | XP_008767950.1 | 3 - 4 | 603 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmx1a
Genomic Map