Assay Details
| Gene Symbol: Slc26a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107172.1 | NP_001100642.1 | 12 - 13 | 1502 | 73 | |
| XM_008769466.1 | XP_008767688.1 | 12 - 13 | 1670 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
anion transport
chloride transport
regulation of pH
positive regulation of gene expression
bicarbonate transport
ion transmembrane transport
anion transmembrane transport
sulfate transmembrane transport
chloride transmembrane transport
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
anion:anion antiporter activity
ATPase binding
anion transport
chloride transport
regulation of pH
positive regulation of gene expression
bicarbonate transport
ion transmembrane transport
anion transmembrane transport
sulfate transmembrane transport
chloride transmembrane transport
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
anion:anion antiporter activity
ATPase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031481 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145867 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031476 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031481 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031481 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031476 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031481 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031476 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10763842 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763833 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763841 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763838 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763832 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763836 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763828 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763831 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763844 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763843 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763847 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763834 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763830 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763835 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763849 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763848 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763837 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763829 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763846 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763845 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763840 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10763839 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10763827 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300001379.rn.1 | TC1300000239.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10763827 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21578902 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2038906 | - | - |

Slc26a9
Genomic Map