Assay Details
| Gene Symbol: Tp53i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113775.1 | NP_001107247.1 | 1 - 2 | 116 | 88 | |
| XM_006250040.2 | XP_006250102.1 | 2 - 3 | 899 | 88 | ||
| XM_008769617.1 | XP_008767839.1 | 1 - 2 | 984 | 88 | ||
| GenBank mRNA | BC158771.1 | - | 2 - 3 | 163 | 7124147 | 88 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001113775.1 transcripts.
Set Membership: |
|

Tp53i3
Genomic Map