Assay Details
| Gene Symbol: Tp53i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113775.1 | NP_001107247.1 | 4 - 5 | 340 | 66 | |
| XM_006250040.2 | XP_006250102.1 | 5 - 6 | 1123 | 66 | ||
| XM_008769617.1 | XP_008767839.1 | 4 - 5 | 1208 | 66 | ||
| XM_008769618.1 | XP_008767840.1 | 2 - 3 | 1051 | 66 | ||
| XM_008769619.1 | XP_008767841.1 | 4 - 5 | 371 | 66 | ||
| XM_008769620.1 | XP_008767842.1 | 4 - 5 | 287 | 66 | ||
| GenBank mRNA | BC158771.1 | - | 5 - 6 | 387 | 7124147 | 66 |
More Information
Set Membership: |
|

Tp53i3
Genomic Map