Assay Details
| Gene Symbol: Tp53i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113775.1 | NP_001107247.1 | 6 - 7 | 531 | 79 | |
| XM_006250040.2 | XP_006250102.1 | 7 - 8 | 1314 | 79 | ||
| XM_008769617.1 | XP_008767839.1 | 6 - 7 | 1399 | 79 | ||
| XM_008769618.1 | XP_008767840.1 | 4 - 5 | 1242 | 79 | ||
| XM_008769619.1 | XP_008767841.1 | 6 - 7 | 562 | 79 | ||
| XM_008769620.1 | XP_008767842.1 | 6 - 7 | 478 | 79 | ||
| XM_008769621.1 | XP_008767843.1 | 3 - 4 | 1177 | 79 | ||
| GenBank mRNA | BC158771.1 | - | 7 - 8 | 578 | 7124147 | 79 |
More Information
Set Membership: |
|

Tp53i3
Genomic Map