Assay Details
| Gene Symbol: Tp53i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113775.1 | NP_001107247.1 | 7 - 8 | 721 | 77 | |
| XM_006250040.2 | XP_006250102.1 | 8 - 9 | 1504 | 77 | ||
| XM_008769617.1 | XP_008767839.1 | 7 - 8 | 1589 | 77 | ||
| XM_008769618.1 | XP_008767840.1 | 5 - 6 | 1432 | 77 | ||
| XM_008769619.1 | XP_008767841.1 | 7 - 8 | 752 | 77 | ||
| XM_008769620.1 | XP_008767842.1 | 7 - 8 | 668 | 77 | ||
| XM_008769621.1 | XP_008767843.1 | 4 - 5 | 1367 | 77 | ||
| GenBank mRNA | BC158771.1 | - | 8 - 9 | 768 | 7124147 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Tp53i3
Genomic Map