Assay Details
| Gene Symbol: LOC103690097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769583.1 | XP_008767805.1 | 465 | 66 | ||
| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047084.1 | NP_001040549.1 | 2 - 3 | 216 | 66 | |
| XM_008769548.1 | XP_008767770.1 | 2 - 3 | 242 | 66 | ||
| GenBank mRNA | AY651776.1 | - | 2 - 3 | 216 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC103690097
Genomic Map