Assay Details
| Gene Symbol: Nmnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048042.1 | NP_001041507.1 | 5 - 6 | 580 | 60 | |
| XM_008769614.1 | XP_008767836.1 | 5 - 6 | 438 | 60 | ||
| GenBank mRNA | DQ022370.1 | - | 5 - 6 | 580 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P020124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120973 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P020124 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P020124 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P020124 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18398 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82171 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33624 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22310 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100401 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58628 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84239 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63166 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86312 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17234 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43732 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42302 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25014 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10764739 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764748 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764746 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764747 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764738 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764742 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764737 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764743 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764740 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764744 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764745 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10764736 | - | - |
| Rat Genome 230 2.0 Array | 1376456_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376456_at | - | - |
| Affymetrix HT Rat Focus Array | 1376456_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376456_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300003461.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003457.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003454.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003453.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003451.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003450.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003460.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003452.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003459.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003449.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003456.rn.1 | TC1300000486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300003455.rn.1 | TC1300000486.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376456_at | - | - |
| Rat Expression Set 230 | 1376456_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10764736 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370377 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353771 | - | - |

Nmnat2
Genomic Map