Assay Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108434.1 | NP_001101904.1 | 3 - 4 | 411 | 58 | |
| XM_006254850.2 | XP_006254912.1 | 3 - 4 | 2665 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
heme biosynthetic process
cholesterol metabolic process
response to light stimulus
detection of UV
response to metal ion
response to lead ion
regulation of gene expression
regulation of eIF2 alpha phosphorylation by heme
response to insecticide
erythrocyte differentiation
very-low-density lipoprotein particle assembly
heme metabolic process
response to drug
response to ethanol
protoporphyrinogen IX metabolic process
response to arsenic-containing substance
regulation of hemoglobin biosynthetic process
response to methylmercury
iron ion homeostasis
response to platinum ion
cellular response to dexamethasone stimulus
ferrochelatase activity
iron ion binding
heme binding
iron-responsive element binding
tetrapyrrole binding
porphyrin-containing compound biosynthetic process
heme biosynthetic process
cholesterol metabolic process
response to light stimulus
detection of UV
response to metal ion
response to lead ion
regulation of gene expression
regulation of eIF2 alpha phosphorylation by heme
response to insecticide
erythrocyte differentiation
very-low-density lipoprotein particle assembly
heme metabolic process
response to drug
response to ethanol
protoporphyrinogen IX metabolic process
response to arsenic-containing substance
regulation of hemoglobin biosynthetic process
response to methylmercury
iron ion homeostasis
response to platinum ion
cellular response to dexamethasone stimulus
ferrochelatase activity
iron ion binding
heme binding
iron-responsive element binding
tetrapyrrole binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P443228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103139 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P443228 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P443228 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P443228 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P443228 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8479 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10804884 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804876 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804878 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804883 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804877 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804886 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804880 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804879 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804885 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804887 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804882 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804888 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10804875 | - | - |
| Rat Genome 230 2.0 Array | 1390388_at | - | - |
| Rat Genome 230 2.0 Array | 1390388_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390388_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390388_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800011117.rn.1 | TC1800001565.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390388_PM_at | - | - |
| Rat Expression Set 230 | 1390388_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10804875 | - | - |
| Rat Genome U34 Set | rc_AI043945_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20798485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040341 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364324 | - | - |

Fech
Genomic Map