Assay Details
| Gene Symbol: Pde6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107386.1 | NP_001100856.1 | 19 - 20 | 2374 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P013666 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P013669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99491 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013669 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013666 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013666 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013669 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P013666 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P013669 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P457179 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62670 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93252 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4238 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10802093 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802106 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802100 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802096 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802099 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802105 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802103 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802104 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802097 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802095 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802102 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802092 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802094 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802098 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10802091 | - | - |
| Rat Genome 230 2.0 Array | 1393426_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393426_at | - | - |
| Affymetrix HT Rat Focus Array | 1393426_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393426_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800004652.rn.1 | TC1800000626.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393426_PM_at | - | - |
| Rat Expression Set 230 | 1393426_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802091 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21591407 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370953 | - | - |

Pde6a
Genomic Map