Assay Details
| Gene Symbol: Zfp474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107380.1 | NP_001100850.1 | 3 - 4 | 400 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107380.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P209220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58073 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P209220 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P209220 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P209220 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P22932 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P209220 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46703 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12263 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10801711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801710 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10801709 | - | - |
| Rat Genome 230 2.0 Array | 1378788_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378788_at | - | - |
| Affymetrix HT Rat Focus Array | 1378788_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378788_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800003715.rn.1 | TC1800000510.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003719.rn.1 | TC1800000510.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003716.rn.1 | TC1800000510.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003718.rn.1 | TC1800000510.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003717.rn.1 | TC1800000510.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800003713.rn.1 | TC1800000510.rn.1 | RTA TC |
| Rat Expression Set 230 | 1378788_at | - | - |
| Rat Expression Set 230 | 1378788_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10801709 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22203856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373341 | - | - |

Zfp474
Genomic Map