Assay Details
| Gene Symbol: Rfx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012172.1 | NP_001012172.1 | 7 - 8 | 872 | 60 | |
| XM_006231233.2 | XP_006231295.1 | 8 - 9 | 1077 | 60 | ||
| XM_006231235.2 | XP_006231297.1 | 7 - 8 | 1036 | 60 | ||
| XM_008760340.1 | XP_008758562.1 | 6 - 7 | 845 | 60 | ||
| XM_008760341.1 | XP_008758563.1 | 7 - 8 | 920 | 60 | ||
| GenBank mRNA | BC085953.1 | - | 7 - 8 | 872 | 7134397 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Rfx3
Genomic Map