Assay Details
Gene Symbol: RGD1305014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014025.1 | NP_001014047.1 | 4 - 5 | 650 | 69 | |
XM_008759885.1 | XP_008758107.1 | 4 - 5 | 1852 | 69 | ||
GenBank mRNA | BC083846.1 | - | 4 - 5 | 650 | 7121351 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
