Assay Details
| Gene Symbol: Slc22a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106327.1 | NP_001099797.1 | 5 - 6 | 928 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a20| Gene Symbol: Slc22a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106327.1 | NP_001099797.1 | 5 - 6 | 928 | 100 | |
Set Membership: |
|