Assay Details
| Gene Symbol: Slc22a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106327.1 | NP_001099797.1 | 6 - 7 | 1155 | 80 | |
| XM_006230750.2 | XP_006230812.1 | 7 - 8 | 1337 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a20
Genomic Map